Learn More
Hach Company Chemical Oxygen Demand (COD) TNTplus Vial Test, ULR (1-60 mg/L COD), 150 Tests
Catalog No. 501998112 Shop All Hach Company Products
Encompass
Hach TNTplus COD vial test reagents are incredibly simple to use and cut down significantly on waste generation compared to other COD test kits. TNT820 is based on Hach Method 10211 for virtually any type of water sample. Hach TNTplus COD vial tests are available in multiple ranges. TNT820 provides a result in the Ultra Low Range of 1 - 60 mg/L COD and is often used for Chemical Oxygen Demand measurement in wastewater effluent. Hach TNT820 does not require a separate blanking vial for further waste reduction. Make the switch to TNTplus COD reagents today!For determination of Chemical Oxygen Demand (COD) by the Reactor Digestion Method. Simply pipette in sample, digest, and read in your Hach Spectrophotometer. 6X25 tests.
Catalog No. 50-199-8112 Supplier Hach Company Supplier No. TNT82006
Hach TNTplus COD vial test reagents are incredibly simple to use and cut down significantly on waste generation compared to other COD test kits. TNT820 is based on Hach Method 10211 for virtually any type of water sample. Hach TNTplus COD vial tests are available in multiple ranges. TNT820 provides a result in the Ultra Low Range of 1 - 60 mg/L COD and is often used for Chemical Oxygen Demand measurement in wastewater effluent. Hach TNT820 does not require a separate blanking vial for further waste reduction. Make the switch to TNTplus COD reagents today!For determination of Chemical Oxygen Demand (COD) by the Reactor Digestion Method. Simply pipette in sample, digest, and read in your Hach Spectrophotometer. 6X25 tests.