Learn More
Hach Company CuVer 1 Copper Reagent Powder Pillows, 10 mL, pk/1000
Supplier:  Hach Company 2105828
Encompass
For copper determination by the bicinchoninate method. EPA compliant for wastewater reporting - Hach Method 8506. Range: up to 5.00 mg/L Cu. Sample Size: 10 mLPack of 1000 powder pillows To confirm accurate results, see appropriate standard solutions on the Consumables tab.
Catalog No. 50-199-5400