Learn More
Hach Company DPD Free Chlorine Reagent Powder Pillows, 10 mL, pk/300
Supplier:  Hach Company 2105503
Encompass
For free chlorine determination by the DPD Free Chlorine method. USEPA accepted for reporting for drinking water analysis using Hach Method 8021. Range: 0.02 to 2.00 mg/L Cl2. Sample Size: 10 mL. Pack of 300 powder pillows.*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-5388