Learn More
Hach Company DPD Total Chlorine Reagent Powder Pillows, 10 mL, pk/300
Catalog No. 501995393 Shop All Hach Company Products
Encompass
For total chlorine determination by the DPD Method. USEPA accepted for reporting for drinking and wastewater analysis using Hach Method 8167. Range: 0.02 - 2.00 mg/L. Sample Size: 10 mL. Pack of 300 powder pillows.Boric Acid free!*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-5393 Supplier Hach Company Supplier No. 2105603
For total chlorine determination by the DPD Method. USEPA accepted for reporting for drinking and wastewater analysis using Hach Method 8167. Range: 0.02 - 2.00 mg/L. Sample Size: 10 mL. Pack of 300 powder pillows.Boric Acid free!*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.