Learn More
Hach Company Filling Solution, ISENa381, 0.02 M NH₄Cl, 59 mL
Catalog No. 501996959 Shop All Hach Company Products
Encompass
Filling solution for Sodium Ion Selective Electrode (ISENa381), Ammonium Chloride (NH₄Cl), 0.02 M, 59 mL bottle
Catalog No. 50-199-6959 Supplier Hach Company Supplier No. 2965126
Filling solution for Sodium Ion Selective Electrode (ISENa381), Ammonium Chloride (NH₄Cl), 0.02 M, 59 mL bottle