Learn More
Hach Company FerroVer Iron Reagent Powder Pillows, 10 mL, pk/1000
Supplier:  Hach Company 2105728
Encompass
For determination of Iron by the 1, 10 phenanthroline method. USEPA approved method for reporting waswater analysis using Hach Method 8008. The FerroVer method recovers total soluble ferrous and ferric iron, as well as complexed iron compounds and many insoluble iron forms. Range: 0.02 to 3.00 mg/L Sample Size: 10 mLPack of 1000 powder pillows To confirm accurate results, see appropriate standard solutions on the Consumables tab.
Catalog No. 50-199-5398