Learn More
Hach Company FerroVer Iron Reagent Powder Pillows, 5 mL, pk/1000
Supplier:  Hach Company 92728
Encompass
For Iron determination by the 1,10 phenanthroline method, using FerroVer Iron Reagent Powder Pillows. For 5-mL samples. Pack of 1000 Powder Pillows
Catalog No. 50-199-4467