Learn More
Hach Company Boron High Range Starter Set
Supplier:  Hach Company 2527900
Encompass
Starter Set for determination of high range boron by the Carmine method. Hach Method 10252. 2 - 50 mg/L BoronSet contains BoroVer 3 Reagent Powder Pillows, Sulfuric Acid, Deionized Water, plus an Erlenmeyer flask, 25 reaction tubes and a cooling rack.
Catalog No. 50-199-6192