Learn More
Hach Company HNB Lab Reagent Package
Supplier:  Hach Company 2307300
Encompass
Each package contains one unit of HNB powder and one unit of HNB dilution solvent. Used for the determination of calcium and magnesium in brine.
Catalog No. 50-199-5762