Learn More
Hach Company HQ1110 Portable Dedicated pH/ORP/mV Meter with Gel ORP Electrode, 5 m Rugged Cable
Catalog No. 502062935 Shop All Hach Company Products
Encompass
HQ1110 Portable pH/ORP Meter, 1 channel for general-purpose field or plant testing with IntelliCAL MTC101 laboratory general purpose ORP electrode, 5 m rugged cable. Field accessories included.
Catalog No. 50-206-2935 Supplier Hach Company Supplier No. LEV015.53.11104
HQ1110 Portable pH/ORP Meter, 1 channel for general-purpose field or plant testing with IntelliCAL MTC101 laboratory general purpose ORP electrode, 5 m rugged cable. Field accessories included.