Learn More
Hach Company HQ1110 Portable Dedicated pH/ORP/mV Meter with Gel pH Electrode, 1 m Cable
Catalog No. 502062936 Shop All Hach Company Products
Encompass
HQ1110 Portable pH/ORP Meter, 1 channel for general-purpose field or plant testing with IntelliCAL PHC201 laboratory general purposes gel filled pH electrode, 1 m cable. Field accessories included.
Catalog No. 50-206-2936 Supplier Hach Company Supplier No. LEV015.53.11101
HQ1110 Portable pH/ORP Meter, 1 channel for general-purpose field or plant testing with IntelliCAL PHC201 laboratory general purposes gel filled pH electrode, 1 m cable. Field accessories included.