Learn More
Hach Company HQ1130 Portable Dedicated Dissolved Oxygen Meter with Dissolved Oxygen Electrode, 5 m Rugged Cable
Catalog No. 502062941 Shop All Hach Company Products
Encompass
HQ1130 Portable DO Meter, 1 channel for aeration basin waste water plant testing with IntelliCAL LDO101 field luminescent/optical dissolved oxygen rugged sensor, 5 m rugged cable. Field accessories included.
Catalog No. 50-206-2941 Supplier Hach Company Supplier No. LEV015.53.11302
HQ1130 Portable DO Meter, 1 channel for aeration basin waste water plant testing with IntelliCAL LDO101 field luminescent/optical dissolved oxygen rugged sensor, 5 m rugged cable. Field accessories included.