Learn More
Hach Company HQ2200 Portable Multi-Meter with pH and Conductivity Electrodes, 1 m Cables
Catalog No. 502062955 Shop All Hach Company Products
Encompass
HQ2200 Portable pH/ORP/EC/TDS/DO Meter, 2 channels for waste water plant testing with IntelliCAL PHC101 gel filled pH and CDC401 Conductivity electrodes, 1 m Cables. Field accessories included.
Catalog No. 50-206-2955 Supplier Hach Company Supplier No. LEV015.53.22001
HQ2200 Portable pH/ORP/EC/TDS/DO Meter, 2 channels for waste water plant testing with IntelliCAL PHC101 gel filled pH and CDC401 Conductivity electrodes, 1 m Cables. Field accessories included.