Learn More
Hach Company HQ4100 Portable Multi-Meter with Conductivity electrode, 5 m Rugged Cable
Catalog No. 502062964 Shop All Hach Company Products
Encompass
HQ4100 Portable pH/ORP/EC/TDS/DO Meter, 1 channel for waste water plant testing with IntelliCAL CDC401 Conductivity electrode, 5 m Rugged Cable. Field accessories included.
Catalog No. 50-206-2964 Supplier Hach Company Supplier No. LEV015.53.41005
HQ4100 Portable pH/ORP/EC/TDS/DO Meter, 1 channel for waste water plant testing with IntelliCAL CDC401 Conductivity electrode, 5 m Rugged Cable. Field accessories included.