Learn More
Hach Company HQ4100 Portable Multi-Meter with gel pH electrode, 5 m Rugged Cable
Catalog No. 502062967 Shop All Hach Company Products
Encompass
HQ4100 Portable pH/ORP/EC/TDS/DO Meter, 1 channel for waste water plant testing with IntelliCAL PHC101 pH electrode, 1 m Rugged Cable. Field accessories included.
Catalog No. 50-206-2967 Supplier Hach Company Supplier No. LEV015.53.41003
HQ4100 Portable pH/ORP/EC/TDS/DO Meter, 1 channel for waste water plant testing with IntelliCAL PHC101 pH electrode, 1 m Rugged Cable. Field accessories included.