Learn More
Hach Company HQ4200 Portable Multi-Meter with Gel pH and Dissolved Oxygen electrode, 1 m Cables
Catalog No. 502062972 Shop All Hach Company Products
Encompass
HQ4200 Portable pH/ORP/EC/TDS/DO Meter, 2 channel for waste water plant testing with IntelliCAL PHC101 pH and CDC401 Conductivity electrode, 5 m Rugged Cables. Field accessories included.
Catalog No. 50-206-2972 Supplier Hach Company Supplier No. LEV015.53.42005
HQ4200 Portable pH/ORP/EC/TDS/DO Meter, 2 channel for waste water plant testing with IntelliCAL PHC101 pH and CDC401 Conductivity electrode, 5 m Rugged Cables. Field accessories included.