Learn More
Hach Company HQ4300 Portable Multi-Meter with Gel pH, Conductivity, and Dissolved Oxygen Electrode, 5 m Rugged Cable
Catalog No. 502062976 Shop All Hach Company Products
Encompass
HQ4300 Portable pH/ORP/EC/TDS/DO Meter, 3 channels for waste water plant testing with IntelliCAL PHC101 pH, CDC401 Conductivity, and LDO101 electrodes, 5 m Rugged Cables. Field accessories included.
Catalog No. 50-206-2976 Supplier Hach Company Supplier No. LEV015.53.43002
Add to Cart
Add to Cart
Request a Quote
HQ4300 Portable pH/ORP/EC/TDS/DO Meter, 3 channels for waste water plant testing with IntelliCAL PHC101 pH, CDC401 Conductivity, and LDO101 electrodes, 5 m Rugged Cables. Field accessories included.