Learn More
Hach Company HQ4300 Portable Multi-Meter with Gel pH, Conductivity, and Dissolved Oxygen Electrode, 1 m Cable
Catalog No. 502062975 Shop All Hach Company Products
Encompass
HQ4300 Portable pH/ORP/EC/TDS/DO Meter, 3 channels for waste water plant testing with IntelliCAL PHC201 pH, CDC401 Conductivity, and LDO101 electrodes, 1 m Cables. Field accessories included.
Catalog No. 50-206-2975 Supplier Hach Company Supplier No. LEV015.53.43001
HQ4300 Portable pH/ORP/EC/TDS/DO Meter, 3 channels for waste water plant testing with IntelliCAL PHC201 pH, CDC401 Conductivity, and LDO101 electrodes, 1 m Cables. Field accessories included.