Learn More
Hach Company HQ440D Water Laboratory Meter Package for Low Ionic Strength Samples with Fast Response RedRod PHC725 Glass Refill Electrode
Catalog No. 501997238 Shop All Hach Company Products
Encompass
Designed for water quality experts, the Hach HQ440D Laboratory Multi-Meter is an advanced laboratory meter that takes the guesswork out of measurements. HQD digital meters connect with smart Intellical electrodes that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time. The advanced HQD Laboratory Meter is designed to withstand years of continuous lab use. HQD bench meters feature a simple user interface that does not require manuals or training to operate. The Intellical PHC725 is a digital, combination glass-body electrode with a refillable KCl reference and built-in temperature sensor. The PHC725 utilizes Red Rod technology to deliver exceptionally fast response times and stable, accurate measurements. It has an annular ring junction and is ideal for measuring pH in a variety of applications, including laboratory measurement of drinking water and low ionic strength (LIS) solutions.
Catalog No. 50-199-7238 Supplier Hach Company Supplier No. 8505850
Designed for water quality experts, the Hach HQ440D Laboratory Multi-Meter is an advanced laboratory meter that takes the guesswork out of measurements. HQD digital meters connect with smart Intellical electrodes that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time. The advanced HQD Laboratory Meter is designed to withstand years of continuous lab use. HQD bench meters feature a simple user interface that does not require manuals or training to operate. The Intellical PHC725 is a digital, combination glass-body electrode with a refillable KCl reference and built-in temperature sensor. The PHC725 utilizes Red Rod technology to deliver exceptionally fast response times and stable, accurate measurements. It has an annular ring junction and is ideal for measuring pH in a variety of applications, including laboratory measurement of drinking water and low ionic strength (LIS) solutions.