Learn More
Hach Company HQ440D Laboratory pH Meter Package for Flat Surface Measurements with Fast Response RedRod PHC729 Glass Refill Electrode
Supplier:  Hach Company 8507450
Encompass
Designed for water quality experts, the Hach HQ440D Laboratory Multi-Meter is an advanced laboratory meter that takes the guesswork out of measurements. HQD digital meters connect with smart Intellical electrodes that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time. The advanced HQD Laboratory Meter is designed to withstand years of continuous lab use. HQD bench meters feature a simple user interface that does not require manuals or training to operate. The Intellical PHC729 utilizes Red Rod technology to deliver exceptionally fast response times and stable, accurate measurements. It has a porous annular ring junction, the flat design makes it ideal for measuring pH on surfaces.
Catalog No. 50-199-7250