Learn More
Hach Company HQ440D Laboratory pH Meter Package for Dirty Samples with Fast Response RedRod PHC735 Glass Refill Electrode
Catalog No. 501997233 Shop All Hach Company Products
Encompass
Designed for water quality experts, the Hach HQ440D Laboratory Multi-Meter is an advanced laboratory meter that takes the guesswork out of measurements. HQD digital meters connect with smart Intellical electrodes that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time. The advanced HQD Laboratory Meter is designed to withstand years of continuous lab use. HQD bench meters feature a simple user interface that does not require manuals or training to operate. The Intellical PHC735 is a digital, combination glass-body electrode with a refillable KCl reference and built-in temperature sensor. The PHC735 utilizes Red Rod technology to deliver exceptionally fast response times and stable, accurate measurements. It has a high-flow annular ring junction and is ideal for measuring pH of “dirty” samples, including wastewater and food and beverage applications.
Catalog No. 50-199-7233 Supplier Hach Company Supplier No. 8505450
Designed for water quality experts, the Hach HQ440D Laboratory Multi-Meter is an advanced laboratory meter that takes the guesswork out of measurements. HQD digital meters connect with smart Intellical electrodes that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time. The advanced HQD Laboratory Meter is designed to withstand years of continuous lab use. HQD bench meters feature a simple user interface that does not require manuals or training to operate. The Intellical PHC735 is a digital, combination glass-body electrode with a refillable KCl reference and built-in temperature sensor. The PHC735 utilizes Red Rod technology to deliver exceptionally fast response times and stable, accurate measurements. It has a high-flow annular ring junction and is ideal for measuring pH of “dirty” samples, including wastewater and food and beverage applications.