Learn More
Hach Company Intellical™ ISENH318101 Ammonia (NH₃) Ion Selective Electrode (ISE) with Calibration Reagents Pack
Catalog No. 501997729 Shop All Hach Company Products
Encompass
The Intellical™ Ammonia ISE Analysis Package contains all you need to start measuring ammonia (NH3) with your HQD meter instrument: The Intellical™ ISENH3181 Ammonia ISE with 1 m cable, 3-pack of replacement Intellical ammonia membrane modules, one 50 mL bottle electrode filling solution, 500 mL bottles of 1 mg/L, 10 mg/L and 100 mg/L of Hach ammonia nitrogen standard solutions, pack of 100 Hach ammonia ionic strength adjustor (ISA) Powder Pillows, a 50 mL bottle of electrode storage solution, and ISE Basic User Manual.
Catalog No. 50-199-7729 Supplier Hach Company Supplier No. ISENH3181AP
The Intellical™ Ammonia ISE Analysis Package contains all you need to start measuring ammonia (NH3) with your HQD meter instrument: The Intellical™ ISENH3181 Ammonia ISE with 1 m cable, 3-pack of replacement Intellical ammonia membrane modules, one 50 mL bottle electrode filling solution, 500 mL bottles of 1 mg/L, 10 mg/L and 100 mg/L of Hach ammonia nitrogen standard solutions, pack of 100 Hach ammonia ionic strength adjustor (ISA) Powder Pillows, a 50 mL bottle of electrode storage solution, and ISE Basic User Manual.