Learn More
Hach Company Intellical™ ISEF12101 Fluoride (F⁻) Ion Selective Electrode (ISE) with Calibration Reagents Pack
Catalog No. 501997723 Shop All Hach Company Products
Encompass
The Intellical™ Fluoride ISE Analysis Package contains all you need to start measuring fluoride (F-) with your HQD meter instrument: The Intellical™ ISEF121 Fluoride ISE with 1 m cable, 500 mL bottles of 0,5 mg/L, 1 mg/L and 2 mg/L of Hach Fluoride standard solutions, pack of 100 Hach fluoride ionic strength adjustor (ISA) Powder Pillows.
Catalog No. 50-199-7723 Supplier Hach Company Supplier No. ISEF121AP
The Intellical™ Fluoride ISE Analysis Package contains all you need to start measuring fluoride (F-) with your HQD meter instrument: The Intellical™ ISEF121 Fluoride ISE with 1 m cable, 500 mL bottles of 0,5 mg/L, 1 mg/L and 2 mg/L of Hach Fluoride standard solutions, pack of 100 Hach fluoride ionic strength adjustor (ISA) Powder Pillows.