Learn More
Hach Company Intellical™ ISENO318101 Nitrate (NO₃⁻) Ion Selective Electrode (ISE) with Calibration Reagents Pack
Catalog No. 501997735 Shop All Hach Company Products
Encompass
The Intellical™ Nitrate ISE Analysis Package contains all you need to start measuring nitrate (NO3-) with your HQD meter instrument: The Intellical™ ISENO3181 Nitrate ISE with 1 m cable, 500 mL bottles of 1 mg/L, 10 mg/L and 100 mg/L of Hach nitrogen-nitrate standard solutions, pack of 100 Hach nitrate ionic strength adjustor (ISA) Powder Pillows.
Catalog No. 50-199-7735 Supplier Hach Company Supplier No. ISENO3181AP
The Intellical™ Nitrate ISE Analysis Package contains all you need to start measuring nitrate (NO3-) with your HQD meter instrument: The Intellical™ ISENO3181 Nitrate ISE with 1 m cable, 500 mL bottles of 1 mg/L, 10 mg/L and 100 mg/L of Hach nitrogen-nitrate standard solutions, pack of 100 Hach nitrate ionic strength adjustor (ISA) Powder Pillows.