Learn More
Hach Company Intellical™ ISENa38101 Sodium (Na⁺) Ion Selective Electrode (ISE) with Calibration Reagents Pack
Catalog No. 501997726 Shop All Hach Company Products
Encompass
The Intellical™ Sodium ISE Analysis Package contains all you need to start measuring Sodium (Na+) with your HQD meter instrument: The Intellical™ ISENA381 Sodium ISE with 1 m cable, the sodium ISAB powder pillows, 100 mg/L and 1000 mg/L sodium standard solutions.
Catalog No. 50-199-7726 Supplier Hach Company Supplier No. ISENA381AP
The Intellical™ Sodium ISE Analysis Package contains all you need to start measuring Sodium (Na+) with your HQD meter instrument: The Intellical™ ISENA381 Sodium ISE with 1 m cable, the sodium ISAB powder pillows, 100 mg/L and 1000 mg/L sodium standard solutions.