Learn More
Hach Company MEL/P-A Safe Drinking Water Lab
Catalog No. 501996251 Shop All Hach Company Products
Encompass
Economical portable lab monitors key water quality parameters such as chlorine (0-3.4 mg/L) and nitrate (0-40 mg/L) plus Total Coliforms and E. Coli (Presence-Absence Method). Includes all reagents for chemistry testing, portable incubator, thermometer, Pocket Pro pH and TDS tester, and glassware - microbiological media sold separately.Please note: PN 2463200 must be ordered separately. This PN is available in the US Only.
Catalog No. 50-199-6251 Supplier Hach Company Supplier No. 2569600
Add to Cart
Add to Cart
Request a Quote
Economical portable lab monitors key water quality parameters such as chlorine (0-3.4 mg/L) and nitrate (0-40 mg/L) plus Total Coliforms and E. Coli (Presence-Absence Method). Includes all reagents for chemistry testing, portable incubator, thermometer, Pocket Pro pH and TDS tester, and glassware - microbiological media sold separately.Please note: PN 2463200 must be ordered separately. This PN is available in the US Only.