Learn More
Hach Company Molybdate Reagent Powder Pillows for HR Silica, 25mL, pk1000
Catalog No. 501994495 Shop All Hach Company Products
Encompass
For determination of high range silica by the Silicomolybdate method. Hach method 8185. Sample size: 25 mL. Pack of 1000 powder pillows
Catalog No. 50-199-4495 Supplier Hach Company Supplier No. 104128
For determination of high range silica by the Silicomolybdate method. Hach method 8185. Sample size: 25 mL. Pack of 1000 powder pillows