Learn More
Hach Company Molybdate Reagent Powder Pillows for HR Silica, 10 mL, pk/1000
Catalog No. 501995417 Shop All Hach Company Products
Encompass
For determination of high range silica by the silicomolybdate method (Hach method 8185) Range:1 to 100 mg/L. Sample Size: 10 mL sample. Pack of 1000 Powder Pillows.
Catalog No. 50-199-5417 Supplier Hach Company Supplier No. 2107328
For determination of high range silica by the silicomolybdate method (Hach method 8185) Range:1 to 100 mg/L. Sample Size: 10 mL sample. Pack of 1000 Powder Pillows.