Learn More
Hach Company NitraVer 5 Nitrate Reagent Powder Pillows, 10 mL, pk/300
Catalog No. 501995405 Shop All Hach Company Products
Encompass
For determination of MR and HR nitrate by the NitraVer 5 cadmium reduction method. 0.1 to 10.0 or 0.3 to 30.0 mg/L as nitrate-N. Hach methods 8039 and 8171. Sample Size: 10 mL. Pack of 300 Powder Pillows
Catalog No. 50-199-5405 Supplier Hach Company Supplier No. 2106103
For determination of MR and HR nitrate by the NitraVer 5 cadmium reduction method. 0.1 to 10.0 or 0.3 to 30.0 mg/L as nitrate-N. Hach methods 8039 and 8171. Sample Size: 10 mL. Pack of 300 Powder Pillows