Learn More
Hach Company NitraVer 5 Nitrate Reagent Powder Pillows, 25 mL, pk/1000
Catalog No. 501994893 Shop All Hach Company Products
Encompass
For mid-range and high range nitrate determination by the NitraVer 5 cadmium reduction method. Hach methods 8039 and 8171. Range: up to 30 mg/L as N. Sample Size: 25 mL. Comes in 10 bags of 100 powder pillows. 1000 total
Catalog No. 50-199-4893 Supplier Hach Company Supplier No. 1403428
For mid-range and high range nitrate determination by the NitraVer 5 cadmium reduction method. Hach methods 8039 and 8171. Range: up to 30 mg/L as N. Sample Size: 25 mL. Comes in 10 bags of 100 powder pillows. 1000 total