Learn More
Hach Company NitriVer 3 Nitrite Reagent Powder Pillows, 25 mL, pk/1000
Catalog No. 501994911 Shop All Hach Company Products
Encompass
For low range nitrite determination by the NitriVer 3 Diazotization Method: EPA Compliant - Hach Method 8507. Range: up to 0.300 mg/L NO2-N Sample Size: 25 mL. Pack of 1000 powder pillows.
Catalog No. 50-199-4911 Supplier Hach Company Supplier No. 1406528
For low range nitrite determination by the NitriVer 3 Diazotization Method: EPA Compliant - Hach Method 8507. Range: up to 0.300 mg/L NO2-N Sample Size: 25 mL. Pack of 1000 powder pillows.