Learn More
Hach Company Buffer Solution Kit, Color-coded, pH 4.01, pH 7.00 and pH 10.01, 20L
Catalog No. 501996112 Shop All Hach Company Products
Encompass
pH Buffer Standard Solution set for pH electrode calibration. Recommended 3-Pack of color-coded pH buffer solutions for easy identification. pH is ±0.02 at 25°C. 20 L Bag-in-box packages of pH 4.01, 7.00 & 10.01, with a collapsible inner bladder in a white corrugated cardboard box. Ideal packaging size for laboratories doing high volume pH testing.
Catalog No. 50-199-6112 Supplier Hach Company Supplier No. 2507300
pH Buffer Standard Solution set for pH electrode calibration. Recommended 3-Pack of color-coded pH buffer solutions for easy identification. pH is ±0.02 at 25°C. 20 L Bag-in-box packages of pH 4.01, 7.00 & 10.01, with a collapsible inner bladder in a white corrugated cardboard box. Ideal packaging size for laboratories doing high volume pH testing.