Learn More
Hach Company Buffer Solution Kit, Color-coded, pH 4.01, pH 7.00 and pH 10.01, 500 mL
Catalog No. 501996934 Shop All Hach Company Products
Encompass
pH Buffer Standard Solution for pH electrode calibration. Pack of 3 color-coded pH buffers. pH is ±0.02 pH units. 500 mL bottles of pH 4.01, 7.00 & 10.01
Catalog No. 50-199-6934 Supplier Hach Company Supplier No. 2947600
pH Buffer Standard Solution for pH electrode calibration. Pack of 3 color-coded pH buffers. pH is ±0.02 pH units. 500 mL bottles of pH 4.01, 7.00 & 10.01