Learn More
Hach Company PhosVer 3 Phosphate Reagent Powder Pillows, 10 mL, pk/1000
Catalog No. 501995402 Shop All Hach Company Products
Encompass
For determination of reactive (ortho) phosphate (phosphorus) by the PhosVer 3 ascorbic acid method. USEPA accepted for reporting for wastewater analysis using Hach Method 8048, Phosphorus, Reactive. Also used in acid-hydrolyzable and total phosphorus determination, after appropriate digestion. Sample Size: 10 mL Pack of 1000 powder pillows. To confirm accurate results, see appropriate standard solutions on the Consumables tab.
Catalog No. 50-199-5402 Supplier Hach Company Supplier No. 2106028
For determination of reactive (ortho) phosphate (phosphorus) by the PhosVer 3 ascorbic acid method. USEPA accepted for reporting for wastewater analysis using Hach Method 8048, Phosphorus, Reactive. Also used in acid-hydrolyzable and total phosphorus determination, after appropriate digestion. Sample Size: 10 mL Pack of 1000 powder pillows. To confirm accurate results, see appropriate standard solutions on the Consumables tab.