Learn More
Hach Company PhosVer 3 Phosphate Reagent Powder Pillows, 25 mL sample, pk/1000
Catalog No. 501994703 Shop All Hach Company Products
Encompass
For phosphate (phosphorus) determination by the PhosVer 3 ascorbic acid method. Hach method 8048. Sample size: 25 mL. Pack of 1000 powder pillows. To confirm accurate results, see appropriate standard solutions on the Consumables tab.
Catalog No. 50-199-4703 Supplier Hach Company Supplier No. 212528
For phosphate (phosphorus) determination by the PhosVer 3 ascorbic acid method. Hach method 8048. Sample size: 25 mL. Pack of 1000 powder pillows. To confirm accurate results, see appropriate standard solutions on the Consumables tab.