Learn More
Hach Company Variable Volume Pipette Set, 1 each 0.2-1.0 mL (BBP078) and 1.0-5.0 mL (BBP065), with tips
Catalog No. 501997889 Shop All Hach Company Products
Encompass
Hach's new variable volume single-channel, air-displacement pipets feature robust design, comfortable ergonomics, slim-line tip ejection, and single-handed operation. Outstanding accuracy with adjustments in 50 μL increments (0.05 mL). Ideal for use with HACH TNTplus reagents. Supplied with calibration report and maintenance tool.
Catalog No. 50-199-7889 Supplier Hach Company Supplier No. LZP320
Hach's new variable volume single-channel, air-displacement pipets feature robust design, comfortable ergonomics, slim-line tip ejection, and single-handed operation. Outstanding accuracy with adjustments in 50 μL increments (0.05 mL). Ideal for use with HACH TNTplus reagents. Supplied with calibration report and maintenance tool.