Learn More
Hach Company Chloride Reagent Set, Mercuric Thiocyanate
Supplier:  Hach Company 2319800
Encompass
For Chloride determination by the Mercuric Thiocyanate method. Hach Method 8113. Range: 0.1 - 25.0 mg/L Cl-. Sample Size: 10 mL. Reagent Set includes Ferric Ion Solution and Mercuric Thiocyanate Solution for 100 tests.
Catalog No. 50-199-5794