Learn More
Hach Company Nickel Reagent Set, Heptoxime
Catalog No. 501995621 Shop All Hach Company Products
Encompass
For determination of nickel by the Heptoxime Method. USEPA accepted method for reporting wastewater analysis results. Hach Method 8037. Range: 0.02 - 1.80mg/L Ni. Set includes Chloroform, Nickel 1 Reagent Powder Pillows and Nickel 2 Reagent Powder Pillows for 50 tests.
Catalog No. 50-199-5621 Supplier Hach Company Supplier No. 2243500
For determination of nickel by the Heptoxime Method. USEPA accepted method for reporting wastewater analysis results. Hach Method 8037. Range: 0.02 - 1.80mg/L Ni. Set includes Chloroform, Nickel 1 Reagent Powder Pillows and Nickel 2 Reagent Powder Pillows for 50 tests.