Learn More
Hach Company Lead Reagent Set, Dithizone Method, (No Air Shipments)
Supplier:  Hach Company 2243100
Encompass
For determination of lead by the Dithizone extraction method. USEPA accepted for reporting for wastewater analysis using Hach Method 8033. Range: 3 to 300 µg/L Pb. Set Includes reagents for 100 tests. For air freight shipments, order 2243101
Catalog No. 50-199-5616