Learn More
Hach Company Manganese Reagent Set, LR, PAN, 10 mL
Supplier:  Hach Company 2651700
Encompass
For determination of Low Range Manganese by the PAN method. Hach Method 8149. Range: 0.006 - 0.700 mg/L Mn. Sample Size: 10 mL. Reagent set includes Alkaline Cyanide Reagent, Ascorbic Acid Powder Pillows and PAN Indicator Solution (0.1%) for approximately 50 tests.
Catalog No. 50-199-6438