Learn More
Hach Company Manganese Reagent Set, LR, PAN, 25 mL
Supplier:  Hach Company 2243300
Encompass
For determination of Manganese by the PAN Method. Hach Method 8149. Range: 0.006 - 0.700 mg/l Mn. Sample Size: 25 mL. Reagent Set includes: Ascorbic Acid Powder Pillows, Alkaline-Cyanide Reagent and PAN Indicator Solution (0.1%) for 100 tests.
Catalog No. 50-199-5619