Learn More
Hach Company Chlorine (Free and Total) Reagent Set, DPD FEAS, Digital Titrator
Catalog No. 501996012 Shop All Hach Company Products
Encompass
For determination of Chlorine (Free and Total) by Ferrous Ethylenediammonium Sulfate Digital Titration, using DPD indicator. Range: Up to 3.00 mg/L Cl2. Reagent Set includes: DPD Free Chlorine Reagent Powder Pillows, DPD Total Chlorine Reagent Powder Pillows, Ferrous Ethylenediammonium Sulfate Titration Cartridge, 0.00564 N. Approximately 40-100 tests.Boric Acid free!*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-6012 Supplier Hach Company Supplier No. 2445300
For determination of Chlorine (Free and Total) by Ferrous Ethylenediammonium Sulfate Digital Titration, using DPD indicator. Range: Up to 3.00 mg/L Cl2. Reagent Set includes: DPD Free Chlorine Reagent Powder Pillows, DPD Total Chlorine Reagent Powder Pillows, Ferrous Ethylenediammonium Sulfate Titration Cartridge, 0.00564 N. Approximately 40-100 tests.Boric Acid free!*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.