Learn More
Hach Company Hypochlorite (Bleach) Reagent Set (HR), for Digital Titrator
Catalog No. 501996544 Shop All Hach Company Products
Encompass
For determination of Hypochlorite (Bleach) by Digital Titrator titration. Hach Method 10100. Range: 5 - 15% Cl2 (50 - 150 g/L Cl2). Stabilized sodium thiosulfate formulation.
Catalog No. 50-199-6544 Supplier Hach Company Supplier No. 2687000
For determination of Hypochlorite (Bleach) by Digital Titrator titration. Hach Method 10100. Range: 5 - 15% Cl2 (50 - 150 g/L Cl2). Stabilized sodium thiosulfate formulation.