Learn More
Hach Company Iron (Total) Reagent Set, HR, Digital Titrator
Catalog No. 501996031 Shop All Hach Company Products
Encompass
For determination of iron by EDTA Digital Titrator titration. Hach Method 8214. Range: 100 - 1000 mg/L as Fe. Reagent Set Contains: Citrate Buffer Powder Pillows, Sodium Periodate Powder Pillows, Sulfosalicylic Acid Powder Pillows and 0.716 M EDTA Digital Titrator Cartridge. Approximately 40-100 tests.
Catalog No. 50-199-6031 Supplier Hach Company Supplier No. 2449300
For determination of iron by EDTA Digital Titrator titration. Hach Method 8214. Range: 100 - 1000 mg/L as Fe. Reagent Set Contains: Citrate Buffer Powder Pillows, Sodium Periodate Powder Pillows, Sulfosalicylic Acid Powder Pillows and 0.716 M EDTA Digital Titrator Cartridge. Approximately 40-100 tests.