Learn More
Hach Company Total Trihalomethanes (THM) Reagent Set
Supplier:  Hach Company 2790800
Encompass
For determination of Total Trihalomethanes by the THM Plus™ Method. Hach Method 10132. Range: 10-600 μg/L CHCl3. Reagent Set includes: THM Plus™ Reagent 1, 2, 3 Solutions and THM Plus 4 Powder Pillows for 50-99 tests. Use as a screening tool to optimize treatment processes, to locate areas of high THM levels in distribution systems and to determine levels of trihalomethanes in drinking water samples.
Catalog No. 50-199-6677