Learn More
Hach Company Silica Reagent Set, Ultra Low Range Rapid Liquid™
Supplier:  Hach Company 2678500
Encompass
For determination of ultra-low range Silica by the Heteropoly Blue Rapid Liquid™ method. Hach method 8282.Range: 3 - 1000 μg/L Set includes: Molybdate 3 Reagent Solution, Citric Acid F Reagent Solution, Amino Acid Reagent Dilution Solvent and Amino Acid F Reagent Powder for approximately 250 tests.Requires Pour-Thru Cell.(Customer mixes powder and diluent upon receipt)
Catalog No. 50-199-6535