Learn More
Hach Company Detergents Test Kit, Model DE-2
Supplier:  Hach Company 143203
Encompass
The presence of detergents in surface or ground waters indicates industrial or domestic pollution. In natural waters anionic LAS and ABS detergents cause excessive foaming, produce undesirable taste and odors, promote growth of disease-producing organisms, and tend to kill aquatic life.
Catalog No. 50-199-4558