Learn More
Hach Company Triazole Test Kit, Model TZ-1, with 230 Vac UV Lamp and Power Supply
Catalog No. 501995495 Shop All Hach Company Products
Encompass
Proper maintenance and economy of operation of modern boilers and cooling towers require a careful balance between water quality parameters and treatment chemicals. Utilizing the color disc method, this kit contains 50 tests.
Catalog No. 50-199-5495 Supplier Hach Company Supplier No. 2167502
Proper maintenance and economy of operation of modern boilers and cooling towers require a careful balance between water quality parameters and treatment chemicals. Utilizing the color disc method, this kit contains 50 tests.