Learn More
Wilmad Labglass Cylindrical Reaction Kettle 1000mL
Catalog No. 50806974
Encompass_Preferred
Upper end is tooled to obtain a thickened portion and flat top. Top is finely ground for a leak-proof fit with covers LG-8030 to LG-8050, ML-1285 and ML-1286.
Catalog No. 50-806-974 Supplier Wilmad Labglass Supplier No. LG8010102
Upper end is tooled to obtain a thickened portion and flat top. Top is finely ground for a leak-proof fit with covers LG-8030 to LG-8050, ML-1285 and ML-1286.