Learn More
Wilmad Labglass Cylindrical Reaction Vessel; O-Ring Flange 3000L
Catalog No. 50805855
Encompass_Preferred
Reaction vessel has a tooled flat flange with an O-ring groove for a greaseless seal. Suitable for vacuum operation. Wall thickness is uniform throughout flask.
Catalog No. 50-805-855 Supplier Wilmad Labglass Supplier No. LG8075102
Reaction vessel has a tooled flat flange with an O-ring groove for a greaseless seal. Suitable for vacuum operation. Wall thickness is uniform throughout flask.